YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
Manufacturer: YSI™ 066155
 View more versions of this product
Catalog No. 13-298-82
  Check Availability
Add to Cart
    Specifications
    Probe Type DO
    Includes 1 mil. Teflon cap, electrolyte solution
    Type Membrane Kit
    For Use With Replenishing Oxygen sensor on YSI™ DO Probe
  • Description & Specifications
Related Products
Product Content Feedback