Learn More
YSI™ ProODO Optical DO Cable, 60m
For use with ProODO, ProDSS, and 5500D instruments.
Manufacturer:  YSI™ 62625060
Catalog No. 14-660-213
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • ODO Digital probe assembly with cable
  • Includes ODO optical DO sensor, sensing cap, cable management kit
  • For ODO and temperature
  • Cable Length: 60m
Specifications
Specifications
ProODO Optical DO Cable
ODO optical DO sensor, sensing cap, cable management kit