Learn More
YSI™ ProODO Handheld Optical Dissolved Oxygen Meter Kit
Provides extreme durability for measurement of optical-based dissolved oxygen for any field application
Manufacturer:  YSI™ 603197
Catalog No. 09-324-507
Add to cart
Includes: ProODO meter, Lab dock, US power supply and ProComm II power, BOD Analyst Pro desktop software for BOD calculations, manual, batteries, communications saddle, USB cable
Description
Description
The ProODO instrument meets the demands of true field work.
  • No electrodes to clean or solutions to change
  • Easy to read graphic, backlit display and keypad for use in any lighting condition
  • Expanded DO range of 0-500% with a response time of T90 in approximately 25 seconds
  • Stores 5000 data sets (sensor data, date, time, site and user defined information)
  • USB connectivity for use with Data Manager software
  • Non-consumptive luminescent method eliminates stirring
  • Impact resistant, IP-67 waterproof case and connectors with rubber non-slip case
  • User replaceable sensing element good for approximately one year
NOTE: Probes and cables are not included and must be purchased separately.
Warranty and Services
Three-year instrument warranty
Specifications
Specifications
Pro ODO Lab Kit, US Pro
ProODO meter, Lab dock, US power supply and ProComm II power, BOD Analyst Pro desktop software for BOD calculations, manual, batteries, communications saddle, USB cable
0.1°C
80 hr.
67
1.05 lb.
0 to 20mg/L, ±0.1mg/L or ±1% of reading, whichever is greater, 20 to 50mg/L, ±10% of the reading
21.6cm
0 to 70ppt
0 to 500% Air Saturation
23° to 158°F
3 Years
Optical
±0.2°C
C Alkaline
RoHS, CE, WEEE, C-Tick, VCCI, FCC, IP67, GLP
0.47kg
2.21 in.
0 to 50mg/L
0.01, 0.1mg/L
0.1% Air Saturation
3.25 in.