Learn More
YSI™ ProODO Handheld Optical Dissolved Oxygen Meter
Provides extreme durability for measurement of optical-based dissolved oxygen for any field application
Manufacturer:  YSI™ 626281
Catalog No. 14-660-204
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The ProODO instrument meets the demands of true field work.
  • No electrodes to clean or solutions to change
  • Easy to read graphic, backlit display and keypad for use in any lighting condition
  • Expanded DO range of 0-500% with response time of T90 in approximately 25 seconds
  • Stores 5000 data sets (sensor data, date, time, site and user defined information)
  • USB connectivity for use with Data Manager software
  • Non-consumptive luminescent method eliminates stirring
  • Impact resistant, IP-67 waterproof case and connectors with rubber non-slip case
  • User replaceable sensing element good for approximately one year
NOTE: Probes and cables are not included and must be purchased separately.
Order Info
Meter Kit is also available (09324507).
Warranty and Services
Three-year instrument warranty
Specifications
Specifications
ProODO Meter
Data Manager software, the ProComm II communications saddle, and USB cable
0.1°C
80 hr.
67
1.05 lb.
0 to 20mg/L, ±0.1mg/L or ±1% of reading, whichever is greater, 20 to 50mg/L, ±10% of the reading
21.6cm
0 to 70ppt
0 to 500% Air Saturation
23° to 158°F
3 Years
Optical
±0.2°C
C Alkaline
RoHS, CE, WEEE, C-Tick, VCCI, FCC, IP67, GLP
0.47kg
2.21 in.
0 to 50mg/L
0.01, 0.1mg/L
0.1% Air Saturation
3.25 in.