Learn More
YSI™ ProODO™ Lab Kit
Manufacturer:  YSI™ 603195
 View more versions of this product
Catalog No. 09-324-505
Add to cart
Specifications
Specifications
3 years instrument, 2 years cable and probe and 1 year sensor cap
±1.5mmHg (0° to 50°C)
80 hr.
RoHS, CE, WEEE, C-Tick, VCCI, IP-67, 1-meter drop test, Assembled in USA, GLC
47.5kg
2.21 in.
23° to 158°F
3.25 in.
US Power Supply
±0.2°C
2 alkaline C-cells
1.05 lb.
0 to 20mg/L ±0.1mg/L or ±1% of reading, 20 to 50mg/L ±10% of the reading
5.6cm
-5° to 70°C