Learn More
YSI™ Reagent Replacement Packs for 9300 and 9500 Photometers
Manufacturer:  YSI™ YAP163
 View more versions of this product
Catalog No. 15-399-28R
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
9300 and 9500 Photometer
Reagent replacement pack
0 to 20 mg/L
200 Tests/Pack
Nitrate