YSI™ Reagent Replacement Packs for 9300 and 9500 Photometers
Manufacturer: YSI™ YAP254
 View more versions of this product
Catalog No. 15-399-18R
  Check Availability
Add to Cart
    Specifications
    Analytes Hardness (Hardicol)
    Packaging 250 Tests/Pack
    Range 0 to 500mg/L
    For Use With 9300 and 9500 Photometer
    Type Reagent replacement pack
  • Description & Specifications
Related Products
Product Content Feedback