YSI™ Reagent Starter Kits for 9300 and 9500 Photometers
Manufacturer: YSI™ YPM250
 View more versions of this product
Catalog No. 15-399
  Check Availability
Add to Cart
    Specifications
    Analytes Alkalinity-Methol Orange
    Range 0 to 500mg/L
    For Use With 9300 and 9500 Photometer
    Includes Reagents, instructions, accessories
    Packaging 50 Tests/Pack
    Type Reagent starter kit
  • Description & Specifications
Related Products
Product Content Feedback