Learn More
YSI™ Reagent Starter Kits for 9300 and 9500 Photometers
For 9300 and 9500 Photometers.
Supplier: YSI™ YPM114
Description
- Includes reagents sufficient for 50 tests
Specifications
Phosphate HR | |
Reagents, instructions, accessories | |
0 to 100 mg/L |
9300 and 9500 Photometer | |
50 Tests/Pk. |