YSI™ Self-stirring BOD Probes for Models 5000 and 5100
Plug-in probes for more convenient functioning.
Manufacturer: YSI™ 050113
 View all options for this product
Catalog No. 15-176-20
  Check Availability
Add to Cart
    For Use With Model 5000 and 5100 DO Meter
    Probe Type Self stirring BOD Probe
    Includes 6.75 in. Shaft for use in wine industry, six membrane caps and KCl probe solution
    Warranty 1 Year
    Model 5010 Self-stirring BOD Probes
    • Plug directly into Models 5000 and 5100 (no adapter cable needed)
    • Operate from meter's power supply
    • Built-in stirrer creates consistent flow past membrane for accurate measurement, faster response
    • Designed to for 300mL BOD bottles
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback