Learn More
YSI™ Weight Sensor/Cable for Professional Plus
For use with Professional Plus instruments.
Manufacturer:  YSI™ 063019
Catalog No. 15-178-110
Add to cart
Description
Description
  • For use with 600R, 600XLM, 600XL, All dual port probe assemblies
  • Attaches to the end of the probe guard
  • Ideal for longer cables to ensure you reach the depths you need
  • Measures 7.62cm (3 in.)
  • Weighs: 0.68kg (24 oz.)
Specifications
Specifications
Ideal for longer cables to ensure you reach the depths you need.
3 in.
24 oz.
Weight, Sensor/Cable
600R, 600XLM, 600XL, All dual port probe assemblies
7.62 cm
0.68 kg