Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ytterbium, 99.9%, (trace metal basis), -12mm, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Description
For ultra trace analysis
Specifications
Color | Gray |
Physical Form | Pieces |
Quantity | 5 g |
Assay Percent Range | 99.9% |
Linear Formula | Yb |
Merck Index | 15, 10303 |
Formula Weight | 173.04 |
Grade | Trace Metal Basis |
Packaging | Glass Bottle |
Chemical Name or Material | Ytterbium, 99.9%, -12 mm |
Safety and Handling
Hazard Category | MOISTURE SENSITIVE |
Hazard Statement | GHS H Statement: Harmful if inhaled. Harmful in contact with skin. Causes skin irritation. May cause respiratory irritation. Harmful if swallowed. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 231-173-2 |
RTECSNumber | ZG1925000 |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction