Alfa Aesar™ Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1457418

Quantity 50g

 View more versions of this product

Catalog No. AA1457418

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 526.11 (Anhydrous)
    MDL Number MFCD00050146
    Chemical Name or Material Ytterbium(III) carbonate hydrate
    Grade Reacton™

    Specifications

    Formula Weight 526.11 (Anhydrous)
    Physical Form Crystalline
    Quantity 50g
    TSCA TSCA
    MDL Number MFCD00050146
    Chemical Name or Material Ytterbium(III) carbonate hydrate
    Assay 99.9%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit