Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium(III) chloride hexahydrate, 99.99+%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
10035-01-5
Cl3Yb·6H2O
10035-01-5
Cl3Yb·6H2O
for ultra trace analysis
Assay Percent Range 99.99+%
Linear Formula YbCl3·6H2O
Formula Weight 387.48
Physical Form Crystals
Percent Purity 99.99+%
Packaging Glass bottle
Total Trace Metal Impurities 100ppm max.
Color Brown to Brown-Red
Quantity 10g
Grade For trace metal basis analysis
Chemical Name or Material Ytterbium(III) chloride hexahydrate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.