Ytterbium(III) oxide, 99.998% (REO), Alfa Aesar™, Reacton™

Manufacturer: Alfa Aesar 1091914

Quantity 25g

 View more versions of this product

Catalog No. AA1091914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-37-0
    Physical Form Powder
    Assay Percent Range 99.998% (REO)
    Molecular Formula Yb2O3
    Formula Weight 394.08
    MDL Number MFCD00011290
    Chemical Name or Material Ytterbium(III) oxide

    Specifications

    Formula Weight 394.08
    Quantity 25g
    EINECS Number 215-234-0
    Physical Form Powder
    Density 9.2g/mL
    Assay Percent Range 99.998% (REO)
    Molecular Formula Yb2O3
    MDL Number MFCD00011290
    CAS 1314-37-0
    Chemical Name or Material Ytterbium(III) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit