Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium sputtering target, 50.8mm dia x 3.18mm thick, Thermo Scientific™

Click to view available options
Quantity:
1 ea.
7440-64-4
Yb
173.054
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
23992
CHEBI:33381
ytterbium
[Yb]
7440-64-4
Yb
173.054
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
23992
CHEBI:33381
ytterbium
[Yb]
Quantity 1 ea.
Odor Odorless
Chemical Name or Material Ytterbium sputtering target, 50.8mm dia. x 3.18mm thick

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 231-173-2

TSCA : Yes

Recommended Storage : Ambient temperatures