Alfa Aesar™ Yttrium foil, 0.025mm (0.001 in.) thick, 99.9% (REO)

Manufacturer: Alfa Aesar™ 12443FF

Quantity 25x25mm

 View more versions of this product

Catalog No. AA12443FF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-65-5
    MDL Number MFCD00011468
    UN Number UN3178
    Chemical Name or Material Yttrium foil, 0.025mm (0.001 in.) thick

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    Physical Form Foil
    MDL Number MFCD00011468
    UN Number UN3178
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    Weight ≈0.07g/25x25mm
    TSCA TSCA
    CAS 7440-65-5
    Odor Odorless
    Storage Note 1
    • Packaged under argon
    Quantity 25x25mm
    Chemical Name or Material Yttrium foil, 0.025mm (0.001 in.) thick
    Assay 99.9%, (REO)

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.

    • Packaged under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit