Alfa Aesar™ Yttrium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1315422

Quantity 100g

 View more versions of this product

Catalog No. AA1315422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 207801-29-4
    Formula Weight 386.24 (Anhydrous)
    MDL Number MFCD00013509
    Synonym Yttrium(III) acetylacetonate
    Chemical Name or Material Yttrium(III) 2,4-pentanedionate hydrate

    Specifications

    Formula Weight 386.24 (Anhydrous)
    MDL Number MFCD00013509
    Synonym Yttrium(III) acetylacetonate
    Physical Form Powder
    CAS 207801-29-4
    Quantity 100g
    Chemical Name or Material Yttrium(III) 2,4-pentanedionate hydrate
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit