Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 3666106

Quantity 5g

 View more versions of this product

Catalog No. AA3666106


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

103470-68-4
MFCD00070441
Yttrium(III) 2-ethylhexanoate
518.16
2-Ethylhexanoic acid yttrium(III) salt

Specifications

5g
Powder
MFCD00070441
Yttrium(III) 2-ethylhexanoate
103470-68-4
518.16
2-Ethylhexanoic acid yttrium(III) salt
99.8%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

GHS P Statement
P280h
Wear protective gloves/clothing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit