Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Yttrium(III) 2-methoxyethoxide, 5% w/v in 2-methoxyethanol

Manufacturer:  Alfa Aesar™ 41793AE

Quantity 100mL

 View more versions of this product

Catalog No. AA41793AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

115668-57-0
314.165
UN1188
Y(OCH2CH2OCH3)3
MFCD00156527
Yttrium(III) 2-methoxyethoxide

Specifications

5% w/v in 2-methoxyethanol
100mL
Liquid
314.165
UN1188
Moisture Sensitive
115668-57-0
Y(OCH2CH2OCH3)3
MFCD00156527
Yttrium(III) 2-methoxyethoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit