Alfa Aesar™ Yttrium(III) 2-methoxyethoxide, 5% w/v in 2-methoxyethanol

Manufacturer: Alfa Aesar™ 41793AC

Quantity 25mL

 View more versions of this product

Catalog No. AA41793AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 115668-57-0
    Linear Formula Y(OCH2CH2OCH3)3
    Formula Weight 314.165
    MDL Number MFCD00156527
    UN Number UN1188
    Chemical Name or Material Yttrium(III) 2-methoxyethoxide

    Specifications

    Formula Weight 314.165
    Quantity 25mL
    CAS 115668-57-0
    Health Hazard 3 GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Physical Form Liquid
    MDL Number MFCD00156527
    UN Number UN1188
    Health Hazard 1 Danger
    Sensitivity Moisture Sensitive
    Linear Formula Y(OCH2CH2OCH3)3
    Health Hazard 2 GHS H Statement
    H360-H226-H302-H312-H332
    May damage fertility or the unborn child.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
    Concentration 5% w/v in 2-methoxyethanol
    Chemical Name or Material Yttrium(III) 2-methoxyethoxide

    GHS H Statement
    H360-H226-H302-H312-H332
    May damage fertility or the unborn child.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P201-P308+P313
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit