Alfa Aesar™ Yttrium(III) acetate tetrahydrate, 99.9% (REO)

In analytical chemistry

Manufacturer: Alfa Aesar™ 1456530

Quantity 250g

 View more versions of this product

Catalog No. AA1456530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85949-60-6
    Linear Formula Y(OOCCH3)3·4H2O
    Formula Weight 338.10 (266.04 Anhydrous)
    MDL Number MFCD00013048
    Chemical Name or Material Yttrium(III) acetate tetrahydrate

    Specifications

    Formula Weight 338.10 (266.04 Anhydrous)
    TSCA TSCA
    MDL Number MFCD00013048
    Quantity 250g
    Solubility Information Soluble in hot water
    Color White
    Density 1.5g/mL
    Physical Form Powder
    Linear Formula Y(OOCCH3)3·4H2O
    CAS 85949-60-6
    Chemical Name or Material Yttrium(III) acetate tetrahydrate
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit