Yttrium(III) Chloride, Anhydrous, 99.9% (REO), Alfa Aesar™

Manufacturer: Alfa Aesar 1868230

Quantity 250g

 View more versions of this product

Catalog No. AA1868230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10361-92-9
    Physical Form -20 Mesh
    Assay Percent Range 99.9%
    Molecular Formula Cl3Y
    Formula Weight 195.26
    MDL Number MFCD00011470
    Chemical Name or Material Yttrium(III) Chloride, Rare Earth Oxides Content

    Specifications

    Formula Weight 195.26
    Quantity 250g
    EINECS Number 233-801-0
    Physical Form -20 Mesh
    Anhydrous or Hydrate Anhydrous
    Assay Percent Range 99.9%
    Molecular Formula Cl3Y
    MDL Number MFCD00011470
    CAS 10361-92-9
    Health Hazard 1 Irritating to eyes, respiratory system and skin.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
    Chemical Name or Material Yttrium(III) Chloride, Rare Earth Oxides Content

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

    Irritating to eyes, respiratory system and skin.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit