Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
10025-94-2
Cl36H2O
303.35
MFCD00149941
10025-94-2
Cl36H2O
303.35
MFCD00149941
for ultra trace analysis
Assay Percent Range 99.90%
Linear Formula YCl3·6H2O
Merck Index 14, 10107
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 303.35
Physical Form Adhering Crystals or Crystalline Powder
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohols
Total Trace Metal Impurities 0.1% max.
Color White
Melting Point 100.0°C
Quantity 50g
Chemical Name or Material Yttrium(III) chloride hexahydrate, 99.9%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 233-801-0

RTECSNumber : ZG3150000

TSCA : TSCA