Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) chloride hexahydrate, 99.999%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
10025-94-2
Cl3Y·6H2O
MFCD00149941
10025-94-2
Cl3Y·6H2O
MFCD00149941
for ultra trace analysis
Assay Percent Range 100.00%
Linear Formula YCl3·6H2O
Merck Index 14, 10107
Formula Weight 303.35
Physical Form Adhering Crystalline Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass Bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohols
Color White
Melting Point 100.0°C
Quantity 10g
Chemical Name or Material Yttrium(III) chloride hexahydrate, 99.999%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-801-0

RTECSNumber : ZG3150000

TSCA : TSCA