Alfa Aesar™ Yttrium(III) chloride hydrate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1118414

Quantity 25g

 View more versions of this product

Catalog No. AA1118414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12741-05-8
    Formula Weight 195.26 (Anhydrous)
    MDL Number MFCD00149941
    Merck Index 14,10107
    Chemical Name or Material Yttrium(III) chloride hydrate
    Grade Reacton™

    Specifications

    Formula Weight 195.26 (Anhydrous)
    Odor Odorless
    Physical Form Wet Crystalline Aggregates
    CAS 12741-05-8
    TSCA TSCA
    Sensitivity Hygroscopic
    Merck Index 14,10107
    Quantity 25g
    MDL Number MFCD00149941
    Chemical Name or Material Yttrium(III) chloride hydrate
    Assay 99.9%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit