Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Yttrium(III) isopropoxide, 20-25% w/w in toluene

Manufacturer:  Alfa Aesar™ 14524NH

Quantity ≈1g

 View more versions of this product

Catalog No. AA14524NH


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2172-12-5
266.17
UN3274
Y[OCH(CH3)2]3
MFCD00015641
Yttrium(III) isopropoxide

Specifications

20 to 25% w/w in toluene
Aromatic
Yellow to brown
2172-12-5
Y[OCH(CH3)2]3
MFCD00015641
Yttrium(III) isopropoxide
0.955g/mL
Air & Moisture Sensitive
≈1g
Liquid
266.17
UN3274

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit