Yttrium(III) nitrate hexahydrate, 99.9% (REO), Alfa Aesar™

Manufacturer: Alfa Aesar 1289822

Quantity 100g

 View more versions of this product

Catalog No. AA1289822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13494-98-9
    Physical Form Crystalline
    Assay Percent Range 99.9% (REO)
    Molecular Formula Y(NO3)3·6H2O
    Formula Weight 383.01 (274.92 Anhydrous)
    MDL Number MFCD00149940
    Chemical Name or Material Yttrium(III) nitrate hexahydrate

    Specifications

    Formula Weight 383.01 (274.92 Anhydrous)
    Quantity 100g
    EINECS Number 233-802-6
    Physical Form Crystalline
    Assay Percent Range 99.9% (REO)
    MDL Number MFCD00149940
    CAS 13494-98-9
    Molecular Formula Y(NO3)3·6H2O
    Chemical Name or Material Yttrium(III) nitrate hexahydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit