Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) oxide, 99.999% (REO), REacton™, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
1kg
1314-36-9
Y2O3
MFCD00011473
1314-36-9
Y2O3
MFCD00011473
Assay Percent Range 99.999% (REO)
Solubility Information Insoluble in water. Soluble in alcohol dilute acids
Formula Weight 225.81
Physical Form ≪10 Micron Powder
Density 5.01
Odor Odorless
Color White
Melting Point 2410°C
Quantity 1kg

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 215-233-5

RTECSNumber : ZG3850000

TSCA : Yes

Recommended Storage : Ambient temperatures