Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) oxide, 99.999%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
2g
10g
50g
Packaging:
Glass bottle
1314-36-9
O3Y2
225.81
MFCD00011473
1314-36-9
O3Y2
225.81
MFCD00011473
for ultra trace analysis
Assay Percent Range 99.999% (Y2O3) On TREO base
Linear Formula Y2O3
Merck Index 14, 10107
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute acids
Formula Weight 225.81
Physical Form Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in dilute acids
Color White
Melting Point 2410.0°C
Quantity 2g
Chemical Name or Material Yttrium(III) oxide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-233-5