Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)

Click to view available options
Quantity:
1 ea.

Magnetron sputtering target

MDL Number MFCD00011473
Formula Weight 225.81
Percent Purity 99.99%
Quantity 1 ea.
Chemical Name or Material Yttrium(III) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay (metals basis)

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 215-233-5

TSCA : Yes

Recommended Storage : Ambient temperatures