Alfa Aesar™ Yttrium(III) perchlorate, 50% w/w aq. soln., Reagent Grade

Manufacturer: Alfa Aesar™ 40580NB

Quantity 5g

 View more versions of this product

Catalog No. AA40580NB

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 14017-56-2
    Formula Weight 495.34 (387.25 Anhydrous)
    MDL Number MFCD00150430
    UN Number UN3211
    Chemical Name or Material Yttrium(III) perchlorate
    Grade Reagent Grade

    Specifications

    Formula Weight 495.34 (387.25 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 14017-56-2
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H314
    May intensify fire
    oxidizer.
    Causes severe skin burns and eye damage.
    Physical Form Liquid
    MDL Number MFCD00150430
    UN Number UN3211
    Quantity 5g
    Color Colorless
    Concentration 50% w/w aq. soln.
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
    Chemical Name or Material Yttrium(III) perchlorate
    Grade Reagent Grade

    GHS H Statement
    H272-H314
    May intensify fire
    oxidizer.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit