Learn More
Yttrium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
Yttrium(III) sulfate octahydrate is used as a laboratory reagent, colorant, structural ceramics, optical glasses and catalysts. It finds application in electrical components and photo-optical materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Yellow |
Physical Form | Crystalline |
Quantity | 250 g |
Merck Index | 14,10107 |
Solubility Information | Water solubility decreases with increasing temperature; Soluble in water. |
Formula Weight | 610.11 (465.99 Anhydrous) |
Percent Purity | 99.9% |
Grade | Reacton™ |
Odor | Odorless |
Assay | (REO) |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 236-844-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.