Alfa Aesar™ Yttrium tris(trifluoromethylsulfonyl)imide

Manufacturer: Alfa Aesar™ 4719306

Quantity 5g

 View more versions of this product

Catalog No. AA4719306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 189114-86-1
    Linear Formula [(CF3SO2)2N]3Y
    Formula Weight 929.31
    MDL Number MFCD23380178
    UN Number UN3261
    Synonym Yttrium tris(trifluoromethanesulfonyl)imide; Yttrium tris(trifluoromethanesulfonimide)
    Chemical Name or Material Yttrium tris(trifluoromethylsulfonyl)imide

    Specifications

    Formula Weight 929.31
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    MDL Number MFCD23380178
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Sensitivity Hygroscopic
    Linear Formula [(CF3SO2)2N]3Y
    CAS 189114-86-1
    UN Number UN3261
    Synonym Yttrium tris(trifluoromethanesulfonyl)imide; Yttrium tris(trifluoromethanesulfonimide)
    Color Pale yellow
    Quantity 5g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Yttrium tris(trifluoromethylsulfonyl)imide

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit