Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YUCCA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120779

This item is not returnable.
View return policy
Description
Positive test controls include: Inflorescences, Before inflorescence, Rosette leaf.
YUCCA1 Polyclonal antibody specifically detects YUCCA1 in Plant samples. It is validated for Western BlotSpecifications
YUCCA1 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Plant | |
Antibody | |
IgG |
Western Blot | |
0.9 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Recombinant protein of Arabidopsis thaliana YUCCA1. | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title