Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC74B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549148

Catalog No. PA549148

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

LRRC74B
Polyclonal
PBS with 0.09% sodium azide
Q6ZQY2
Leucine-rich repeat-containing protein 74B, LRRC74B (HGNC:34301)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
400891
Western Blot
Unconjugated
LRRC74B
Liquid
LRRC74B
KLH conjugated synthetic peptide between 355-383 amino acids from the C-terminal region of human YV012.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit