Learn More
Aviva Systems Biology Corp YWHAZ Antibody - C-terminal region (ARP48046_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48046_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against YWHAZ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human YWHAZ. Alternate names: KCIP-1, MGC111427, MGC126532, MGC138156, YWHAD, 14-3-3-zetaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-59709