Learn More
Aviva Systems Biology Corp YWHAZ Antibody - middle region (ARP48047_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48047_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YWHAZ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human YWHAZ. Alternate names: KCIP-1, MGC111427, MGC126532, MGC138156, YWHAD, 14-3-3-zetaApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59710