Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YWHAZ (phospho S58) Rabbit anti-Human, Mouse, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30172
Description
Rabbit polyclonal antibody raised against partial synthetic phosphopeptide of human YWHAZ.
- High specificity
- High reactivity
Specifications
YWHAZ (phospho S58) | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial synthetic phosphopeptide of human YWHAZ. | |
In 1.0 mg/mL PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide). | |
KCIP-1/MGC111427/MGC126532/MGC138156 | |
Rabbit | |
Affinity purification | |
RUO | |
7534 | |
Store at 4°C for short term use. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunofluorescence (1:100-1:200) Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
P63104 | |
YWHAZ | |
A synthetic phosphopeptide corresponding to residues surrounding S58 of human YWHAZ. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction