Learn More
Aviva Systems Biology Corp YY1 Antibody - middle region (P100898_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100898_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YY1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human YY1. Alternate names: DELTA, NF-E1, UCRBP, INO80S, YIN-YANG-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62773