Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YY1 Rabbit anti-Human, Mouse, Rat, Clone: S02-8A3, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314987100UL
This item is not returnable.
View return policy
Description
YY1 Monoclonal antibody specifically detects YY1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
YY1 | |
Monoclonal | |
Unconjugated | |
DELTA, Delta transcription factor, INO80SYY-1, NF-E1INO80 complex subunit S, transcriptional repressor protein YY1, UCRBP, Yin and yang 1, Yin and Yang 1 protein, YIN-YANG-1, YY1 transcription factor | |
A synthetic peptide of human YY1 (Uniprot#: P25490) | |
100 μg | |
Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
7528 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
S02-8A3 | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction