Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ YZ9

PFKFB3 inhibitor

Manufacturer:  Tocris™ 5048/50

 View more versions of this product

Catalog No. 504850R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

PFKFB3 (6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase) inhibitor (IC50 = 0.183µM). Also competitively inhibits F-6-P (Ki = 0.094µM). Inhibits cell growth (GI50 = 2.7µM) in a XTT-based in vitro toxicology assay.
  • Chemical Name: Ethyl 7-hydroxy-2-oxo-2H-1-benzopyran-3-carboxylate
  • Formula: C12H10O5
  • CAS Number: 6093-71-6
  • Solubility: Soluble to 50mM in DMSO and to 20mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

YZ9
>98
50mg
YZ9
234.2
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit