Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Z-Cyclopentyl-AP4, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Group III mGlu receptor agonist that exhibits higher potency at mGlu4 than mGlu8 (EC50 values are 49 and 124 μM respectively) with no activity at mGlu7. Selectively inhibits synaptic activity in the lateral perforant pathway (IC50 values are 130 and 1859 μM in the lateral and medial perforant pathways respectively). Conformationally restrained analog of L-AP4 (Cat No. 0103).
Specifications
Quantity | 50 mg |
Formula Weight | 209.14 |
Percent Purity | >98% |
Chemical Name or Material | Z-Cyclopentyl-AP4 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction