Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(z)-α-fluorocinnamic Acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
20397-61-9
C9H7FO2
166.151
MFCD00004222
QONCEXMULRJPPY-VURMDHGXSA-N
alpha-fluorocinnamic acid,.alpha.-fluorocinnamic acid,cinnamic acid, alpha-fluoro,z-alpha-fluorocinnamic acid,2-propenoic acid, 2-fluoro-3-phenyl,z-2-fluoro-3-phenylacrylic acid,2-fluoro-3-phenyl-2-propenoic acid,2z-2-fluoro-3-phenylprop-2-enoic acid,2-fluoro-3-phenylacrylic acid,cinnamic acid, .alpha.-fluoro
1550911
(Z)-2-fluoro-3-phenylprop-2-enoic acid
C1=CC=C(C=C1)C=C(C(=O)O)F
20397-61-9
C9H7FO2
166.151
MFCD00004222
QONCEXMULRJPPY-VURMDHGXSA-N
alpha-fluorocinnamic acid,.alpha.-fluorocinnamic acid,cinnamic acid, alpha-fluoro,z-alpha-fluorocinnamic acid,2-propenoic acid, 2-fluoro-3-phenyl,z-2-fluoro-3-phenylacrylic acid,2-fluoro-3-phenyl-2-propenoic acid,2z-2-fluoro-3-phenylprop-2-enoic acid,2-fluoro-3-phenylacrylic acid,cinnamic acid, .alpha.-fluoro
1550911
(Z)-2-fluoro-3-phenylprop-2-enoic acid
C1=CC=C(C=C1)C=C(C(=O)O)F
Infrared Spectrum Authentic
Color Brown-Yellow to White
Quantity 5g
Melting Point 118°C to 125°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5CH=C(F)CO2H
Formula Weight 166.15
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material (Z)-α-Fluorocinnamic acid
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger