Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Z-GLY-PRO-ARG-7-AMINO-4-TRIFLUOROMETHYLCOUMARIN

Used to test function of Granzyme A by using an enzymatic in vitro cell free assay.

$403.00 - $832.50

Specifications

Format Solid
Product Type Proteins, Enzymes & Peptides
For Use With (Application) Substrate to test function of Granzyme A
Storage Requirements Keep cool and dry. Desiccate AFC-, AMC-, or MNA061 in solid form at room temperature. Store DMSO/DMF solution at -20 °C. Material is stable for at least one year, if stored as recommended.
Color White
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP3AFC06110 View Documents MP Biomedicals™
03AFC06110
10mg Each for $403.00
Add to cart
 
MP3AFC06125 View Documents MP Biomedicals™
03AFC06125
25mg Each for $832.50
Add to cart
 
Description

Description

Applications: Z-Gly-Pro-Arg-7-Amino-4-Trifluoromethylcoumarin is used to test function of Granzyme A by using an enzymatic in vitro cell free assay. Granzyme A is one of the most abundant granzymes known to be involved in cell death processes, using the alternative programmed cell death pathway.
Product Description: Z-Gly-Pro-Arg-7-Amino-4-Trifluoromethylcoumarin is a fluorescent substrate for granzyme A. Granzyme A is one of the most abundant granzymes known to be involved in cell death processes, using the alternative programmed cell death pathway.
Specifications

Specifications

Solid
Substrate to test function of Granzyme A
White
Proteins, Enzymes & Peptides
Keep cool and dry. Desiccate AFC-, AMC-, or MNA061 in solid form at room temperature. Store DMSO/DMF solution at -20 °C. Material is stable for at least one year, if stored as recommended.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit