Learn More
TARGETMOL CHEMICALS INC ZABEDOSERTIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T370765MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zabedosertib (BAY 1834845) is an inhibitor of IRAK4 with immunomodulatory potential. Zabedosertib protects against acute respiratory distress syndrome a new study suggests. purity: 98%
Catalog No. 50-258-0149