Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149451AP

Catalog No. 50-172-9854


Add to cart

Description

Description

MLKLAK is a member of the MAPKKK family of signal transduction molecules. It possesses an N-terminal kinase catalytic domain, followed by a leucine zipper motif and a sterile-alpha motif (SAM). This magnesium-binding protein forms homodimers and is located in the cytoplasm. The protein mediates gamma radiation signaling leading to cell cycle arrest and activity of this protein plays a role in cell cycle checkpoint regulation in cells. The protein also has pro-apoptotic activity.
Specifications

Specifications

ZAK
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9ESL4, Q9NYL2
AZK, HCCS 4, MLK related kinase, MLK7, mlklak, MLT, MLTK, MRK, ZAK
Rabbit
IgG
150 μL
-20°C
Primary
311743, 51776, 65964
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MAP3K20
Liquid
Map3k20
ZAK Fusion Protein Ag6747
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit