Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ ZAP-70 Ab-1, Mouse Monoclonal Antibody

Provides accurate, reproducible results with the Thermo Scientific™ ZAP-70 Ab-1, Mouse Monoclonal Antibody.

$182.00 - $688.50

Specifications

Antigen ZAP-70 Ab-1
Clone 2F3.2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1911P
View Documents
Thermo Scientific™
MS1911P
Purified Ab with BSA and Azide 1mL Each for $688.50
Add to cart
 
MS1911P0
View Documents
Thermo Scientific™
MS1911P0
Purified Ab with BSA and Azide 100μL Each for $182.00
Add to cart
 
MS1911P1
View Documents
Thermo Scientific™
MS1911P1
Purified Ab with BSA and Azide 500μL Each for $431.00
Add to cart
 
MS1911R7
View Documents
Thermo Scientific™
MS1911R7
Ready-to-use for IHC (UVLP) 7mL Each for $430.50
Add to cart
 
MS1911R1
View Documents
Thermo Scientific™
MS1911R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Provides accurate, reproducible results with the Thermo Scientific™ ZAP-70 Ab-1, Mouse Monoclonal Antibody.

Mutations in ZAP-70 gene results in a form of Severe Combined Immunodeficiency Syndrome (SCID) in humans. ZAP-70 expression also defines a subset of Chronic Lymphocytic Leukaemia (CLL) in patients with unmutated Ig gene and poor clinical course.

Host Species: Mouse

Clone: 2F3.2

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: GST-fusion to tandem SH2 domains of human ZAP-70 corresponding to residues 1-254

Molecular Weight: 70kDa

Positive Control: Tonsil, Jurkat cells, T- and NK-cells

Cellular Localization: Cytoplasmic (with occasional staining of macrophage lysozymes)

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

ZAP-70 Ab-1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IVD
Primary
IgG2a
2F3.2
Monoclonal
Murine
Cancer and Tumor Biology
GST-fusion to tandem SH2 domains of human ZAP-70 corresponding to residues 1-254
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit