Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZAP70 Rabbit anti-Human, Clone: ZAP70/4040R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30722320UG
Description
ZAP70 Monoclonal specifically detects ZAP70 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
ZAP70 | |
Monoclonal | |
0.2 mg/ml | |
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, SRKFLJ17679, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD | |
Synthetic peptide corresponding to residues withinaa200-350 of ZAP70 was used as an immunogen. | |
20 μg | |
Adaptive Immunity, Cell Biology, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
7535 | |
Store at 4°C. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
ZAP70/4040R | |
Unconjugated | |
1 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction