Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZAP70 Rabbit anti-Human, Mouse, Unconjugated, Clone: H11, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19167
Description
ZAP70 Monoclonal antibody specifically detects ZAP70 in Human, Mouse samples. It is validated for WB-Ce, Flow Cyt.Specifications
ZAP70 | |
Monoclonal | |
Unconjugated | |
ZAP70 | |
Rabbit | |
200 μL | |
7535, 6850 | |
Store at -20°C | |
IgG1 κ |
Western Blot, Flow Cytometry | |
H11 | |
zeta-chain (TCR) associated protein kinase 70kDa | |
FLJ17670, FLJ17679, SRK, STD, TZK, ZAP-70 | |
A synthetic phosphopeptide corresponding to residues surrounding Y493 of human ZAP70 | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction